Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Rectangular Buckets
Product Code. 10216842 Shop All Thermo Scientific Products
Click to view available options
Tube Type:
15 mL Blood Collection Tube
15 mL DIN
Conical
Unit Size:
Each
Product Code. 10216842
Brand: Thermo Scientific™ 75002021
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with four-place swinging bucket rotor 75-002-000 when equipped with rectangular bucket 75-002-001
Adapters sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Tube Type 15 mL Blood Collection Tube
For Use With (Equipment) 4-place Swing rotor with rectangle bucket