missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorbitol MacConkey Agar (Dehydrated)

Product Code. p-200112444
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10229192

Brand: Thermo Scientific™ CM0813B

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Selectively detect Escherichia coli O157 with this medium.

Selectively detect Escherichia coli O157 with Thermo Scientific™ Oxoid™ Sorbitol MacConkey Agar (Dehydrated). The formulation, based on that described by Rappaport and Henig1 is identical to MacConkey Agar No.3, except that lactose has been replaced with sorbitol. Escherichia coli O157 does not ferment sorbitol and, therefore, produces colorless colonies. In contrast, most E. coli strains ferment sorbitol and form pink colonies.

Escherichia coli O157 is recognized as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhea and severe abdominal pain, and hemolytic uraemic syndrome (HUS) 2, 3, 4, 5,6, and as such, is a significant human pathogen.

Sorbitol MacConkey Agar is recommended for the isolation of pathogenic E.coli O157.

  • Sensitive and specific for coli O157 based on sorbitol fermentation.
  • Recommended as a simple, inexpensive, rapid and reliable means of screening for E.coli O157

The efficiency of Sorbitol MacConkey Agar has been confirmed by March and Ratnam7. These workers reported a sensitivity of 100% and a specificity of 85% and recommended the medium as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: FDA.
Description Sorbitol MacConkey Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.7L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.