missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sorafenib
GREENER_CHOICE

Product Code. 18785476
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18785476

Brand: Tocris Bioscience™ 6814/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent Raf-1 inhibitor; also inhibits VEGFR-2, VEGFR-3, PDGFR-β, Flt-3 and cKIT

Sorafenib is a potent Raf inhibitor (IC50 values are 6, 22 and 38 nM for Raf-1, wild-type BRAF and V599E BRAF mutant, respectively). Also inhibits mVEGFR-2, mVEGFR-3, mPDGFR-β, Flt-3, cKIT and VEGFR-2 (IC50 values are 15, 20, 57, 58, 68 and 90 nM, respectively). Inhibits MAPK pathway in colon, pancreatic and breast cancer cells expressing mutant KRAS and mutant or wt BRAF in vitro. Exhibits antitumor activity in colon, breast and non small-cell lung cancer xenograft models. Also induces ferroptosis in hepatocellular carcinoma cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide
CAS 284461-73-0
Quantity 10 mg
Molecular Formula C21H16ClF3N4O3
Formula Weight 464.82
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.