Learn More
Syngene™ Sony™ A6 Matte Paper
Product Code. 17776869 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
A6 Digital Black and White Thermal Printer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17776869 A6 Digital Black and White Thermal Printer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17776869 Supplier Syngene™ Supplier No. UPP100HD
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
High density matte print media with proper width for use in the Sony™ A6 Digital Black and White Thermal Printer.
The UP-D898MD is a successor to the popular Sony™ UP-897 series of medical printers. Redesigned and upgraded, it offers several operational and ergonomic enhancements while retaining the same space-saving footprint and easy integration into medical carts.
  • Enhanced usability with enlarged LCD display panel and joystick for intuitive menu navigation
  • Delivers high quality black and white prints in under two seconds
  • Advanced thermal printing technology delivers near-photo quality results using widely available print media
  • Unique Sony™ technology allows paper to be cut in two directions – both up and down – as a lower cutter is newly included, along with the conventional upper cutter
  • Flexible printer installation in almost any location, as well as smooth printing operations
  • High quality thermal print engine can produce hard copy prints of still images captured for use in patient records and referrals
  • Type II matte paper
Specifications
Type Matte Paper
For Use With (Equipment) A6 Digital Black and White Thermal Printer
Dimensions (L x W) 20 m x 110 mm