Learn More
Branson Ultrasonic Sonifier SFX 550 240V CE 3/4" PROBE
Brand:  Branson Ultrasonic 101-063-970R
Sonifier SFX 550 240V CE 3/4" PROBE
Product Code. 15559624
  • ONLINE EXCLUSIVE PRICE
    8641.50€
    6546.00€ / Each
    Save 2095.50€
    24% Off
Only null left
Estimated Shipment: 19-01-2026