Learn More
Fisherbrand™ Q500 Sonicator without Probe
Product Code. 12981151 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Voltage:
220 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
12981151 220 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12981151
  • ONLINE EXCLUSIVE PRICE
    4080.00€
    3195.20€ / Pack of 1
    Save 884.80€
    21% Off
View all promotions
Estimated Shipment: 12-01-2027
Providing programmable operation and a digital display of operating parameters. Fisherbrand™ Q500 Sonicator without Probe is excellent for nanoparticle dispersion, creating emulsions, cell lysis and homogenization applications.
  • Set time and amplitude for hands-free operation
  • Pulse mode to prevent heat-up in temperature-sensitive samples
  • Digital amplitude/intensity control from 20-100%
  • Elapsed time indicator to display duration of sonication
  • Display of wattage and joules for real-time energy monitoring
  • Overload protection to prevent damage to circuitry if a fault occurs
  • Uses lead-free components
Specifications
Voltage 220 V
Wattage 500 W
Dimensions (L x W x H) 387 x 203 x 216 mm (5.25 x 8 x 8.5 in.)
Includes Generator, converter, power cable, converter cable, wrench set
Model Q500
Frequency 20 kHz