Learn More
Cytiva SolVac™ Filter Holder
Description
- For safe and simply mobile phase filtration
- Ensures analysts can obtain the best possible results while effectively protecting their valuable equipment
Specifications
Specifications
| Sterility | Non-sterile |
| Filtration Area | 10.2 cm2 |
| Includes | 61 cm (2 ft.) feed line tubing, thumb clamp, sinker, vacuum port adapter, 2 membrane seal gaskets, and 2 seal gaskets |
| Diameter (Metric) Inside | 47 mm |