Learn More
Memmert™ Software AtmoCONTROL for UF55plus
Product Code. 17569505 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF55plus Oven
Unit Size:
Each
Product Code. 17569505
Brand: Memmert™ UF55plusFDAQ1
Only null left
Add to basket
Add to basket
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Certifications/Compliance Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
For Use With (Equipment) UF55plus Oven
Type Software