Learn More
Memmert™ Software Atmocontrol, Base License For 1 Unit, Iq/Oq Doc. Available In De And En, For Ico240Med
Product Code. 16336887 Shop All Memmert Products
Click to view available options
Includes:
Base License for 1 Unit
Unit Size:
Each
Product Code. 16336887
Brand: Memmert™ ICO240medFDAQ1
Only null left
Add to basket
Add to basket
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base License for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Certifications/Compliance Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
For Use With (Equipment) Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106
Type Software
Includes Base License for 1 Unit