Learn More
Megro™ SOFT-JECT™ Disposable Syringes
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
Unit Size:
Pack of 100
Description
- Larger diameters and higher sealing lips guarantees a better density of the syringe
- Second brake ring plate at the piston rod provides better and exact sliding of the piston rod
- Latex-free
- Clear for excellent optical clarity and visibility of contents
- Piston and cylinder of polypropylene, piston plug of synthetic polyisoprene-rubber
- Luer attachment
Specifications
Specifications
| Material | Polypropylene (Barrel), Polypropylene (Plunger) |
| Color | Transparent |
| For Use With (Application) | Comfortable handling, better accuracy |
| Latex Free | Yes |
| Disposable | Single-use |
| Quantity | 100/Pk. |
| Volume (Metric) | 5 mL |
| Connection Type | Luer |
| Graduated | Yes |