Learn More
Honeywell™ SOFRACOLD GRIP Gloves
Product Code. 12655333
Click to view available options
:
Size: Universal
Product Code. 12655333
Brand: Honeywell™ 2232037
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Honeywell™ SOFRACOLD GRIP Gloves for handling of frozen food or packaging in dry conditions.
  • The knitting technique and Lycra™ elasticity help to obtain a glove that conforms snugly to the hand
  • The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The PVC dots ensure greater abrasion resistance and a better grip
  • The Thermastat™/Lycra™ thread offers excellent protection against the cold, particularly as an underglove
  • The PVC dots ensure greater abrasion resistance and a better grip
  • Blue color makes the glove more soilproof
TRUSTED_SUSTAINABILITY
Specifications
Description 13 gauge
Coating Material Thermastat, Lycra, PVC
Color Blue
Size Universal
Glove Cuff Color Yellow
Length (Metric) 23.5 cm