Learn More
Sodium Ursodeoxycholate, TRC
Product Code. 30415116
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415116 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30415116 Supplier Toronto Research Chemicals Supplier No. TRCS08083325G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium Ursodeoxycholate
CAS 2898-95-5
Molecular Formula C24H39NaO4
Quantity 25 g
Synonym (3alpha,5beta,7beta)-3,7-Dihydroxy-cholan-24-oic acid Monosodium Salt, 3alpha,7beta-Dihydroxy-5beta-Cholan-24-oic Acid Monosodium Salt
InChI Formula InChI=1S/C24H40O4.Na/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26;/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-;/m1./s1
SMILES [Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name sodium;(4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 414.6
Formula Weight 414.2746