missing translation for 'onlineSavingsMsg'
Learn More

Sodium Triflinate, TRC

Product Code. 17964725
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17964725 Supplier Toronto Research Chemicals Supplier No. TRCS6736901G
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2926-29-6
Molecular Formula C F3 O2 S . Na
Quantity 1 g
Synonym Perfluoromethane Sulfinate Sodium, Methanesulfinic acid, 1,1,1-trifluoro-, sodium salt (1:1) (ACI, Methanesulfinic acid, trifluoro-, sodium salt (8CI, 9CI), Langlois reagent, Sodium triflinate, Sodium trifluoromethanesulfinate, Sodium trifluoromethylsulfinate
InChI Formula InChI=1S/CHF3O2S.Na/c2-1(3,4)7(5)6;/h(H,5,6);/q;+1/p-1
SMILES [Na+].[O-]S(=O)C(F)(F)F
IUPAC Name sodium;trifluoromethanesulfinate
Molecular Weight (g/mol) 156.06
Formula Weight 155.9469
Chemical Name or Material Sodium Triflinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.