missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrafluoroborate, 95%

Product Code. 11327287
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy

Product Code. 11327287

Brand: Thermo Scientific Alfa Aesar 011562.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorinating agent | CAS: 13755-29-8 | BF4Na | 109.79 g/mol

Fluorinating agentSodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
IUPAC Name sodium;tetrafluoroborate
SMILES [Na+].F[B-](F)(F)F

Specifications

Melting Point 384°C (decomposition)
Color White
Physical Form Granular/Lump
Quantity 2 kg
Linear Formula NaBF4
Merck Index 14,8617
Solubility Information Soluble in water.
Formula Weight 109.79
Percent Purity 95%
Sensitivity Hygroscopic
Density 2.47 g/mL
Chemical Name or Material Sodium tetrafluoroborate
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.