missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Sodium Test Kits

Product Code. 11767295
Click to view available options
No. of Tests:
25
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11767295

Brand: WTW™ 252044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Analytes Kalignost, turbidometric
For Use With (Application) In nutrient solutions for fertilization
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
No. of Tests 25
Sufficient For 25 Tests
Test Method Photometric
Type Potassium (K) cell test kit
compliance-icons
Product Identifier
  • Sodium Cell Test in nutrient solutions for fertilization
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : iron (III) nitrate,nitric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.