missing translation for 'onlineSavingsMsg'
Learn More

Sodium tellurate(VI) hydrate, Te 45% min

Product Code. p-7080338
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11314889

Brand: Thermo Scientific Alfa Aesar 075106.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10101-83-4 | Na2O4Te | 237.58 g/mol

Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-83-4
Molecular Formula Na2O4Te
Molecular Weight (g/mol) 237.58
MDL Number MFCD00014242
InChI Key XERQTZLDFHNZIC-UHFFFAOYSA-L
Synonym sodium tellurate, unii-7kar02f0r0, natriumtellurat, 2na.o4te, disodium tellurate, telluric acid h2teo4 , sodium salt 1:2, telluric acid h2teo4 ,disodium salt, dihydrate 8ci,9ci
PubChem CID 13652393
SMILES [Na+].[Na+].[O-][Te]([O-])(=O)=O

Specifications

Melting Point (decomposition)
Physical Form Crystalline
Quantity 10 g
Merck Index 14,8686
Solubility Information Soluble in water.
IUPAC Name disodium tellurate
Formula Weight 237.58 (Anhydrous)
Concentration or Composition (by Analyte or Components) Te 45% min
Chemical Name or Material Sodium tellurate (VI) hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.