Learn More
Sodium Selenite, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Sodium Selenite |
| CAS | 10102-18-8 |
| Molecular Formula | 2 Na . O3 Se |
| Quantity | 10 g |
| Synonym | Microgan Se, Disodium Selenium Trioxide, Ekosel C-BB, Neoselen Selamin, Selemag, Sodium Selenium Oxide (Na2SeO3), Ekosel C-BB, Disodium Selenite |
| InChI Formula | InChI=1S/2Na.H2O3Se/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2 |
| SMILES | [Na+].[Na+].[O-][Se](=O)[O-] |
| IUPAC Name | disodium;selenite |
| Molecular Weight (g/mol) | 172.94 |
| Formula Weight | 173.8808 |