missing translation for 'onlineSavingsMsg'
Learn More

Sodium Propionate-d5, TRC

Product Code. 30415580
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415580 100 mg -
30421843 10 mg -
30423911 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30415580 Supplier Toronto Research Chemicals Supplier No. TRCS667066100MG
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202529-18-8
Molecular Formula C3D5NaO2
Molecular Weight (g/mol) 101.09
Synonym Propanoic-d5 acid, sodium salt (9CI), Sodium propionate-D5, Sodium propionate-d5, Sodium propanoate-d5, Propionic acid-d5 sodium salt
IUPAC Name sodium propanoate-d5
SMILES O=C([O-])C([2H])([2H])C([2H])([2H])[2H].[Na+]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1/i1D3,2D2;
Formula Weight 101.05
Chemical Name or Material Sodium Propionate-D5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.