missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdenum oxide dihydrate, 98%

Product Code. 11479496
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11479496

Brand: Thermo Scientific Alfa Aesar A19222.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol

Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

Solubility
Soluble in water.

Notes
Stable. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 100°C (in -2H2O)
Quantity 500 g
Merck Index 14,8645
Solubility Information Soluble in water.
Formula Weight 241.95 (205.92 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 2.71
Chemical Name or Material Sodium molybdenum oxide dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.