missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sodium Metabisulfite, Certified AR for Analysis
Specifications
Specifications
| Melting Point | 150°C |
| Color | White |
| pH | 4 to 6 |
| Physical Form | Powder/Solid |
| Quantity | 1 kg |
| Packaging | Plastic powder jar |
| Formula Weight | 190.1g/mol |
Safety and Handling
Product Identifier
- Sodium metabisulfite
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P233-Keep container tightly closed.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?