missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lauryl Ether Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Lauryl Ether Sulfate |
| CAS | 9004-82-4 |
| Molecular Formula | (C2 H4 O)n C12 H26 O4 S . Na |
| Quantity | 5 g |
| Synonym | Standapol 125EStandapol ES 1Standapol ES 2Standapol ES 3Standapol ES 3DSStandapol ESPSteinapol NL 3Steol 25-3S70FCSteol 4NSteol BES 70Steol CA 370Steol CS 130Steol CS 170Steol CS 230Steol CS 270Steol CS 270CSteol CS 330Steol CS 330LSteol CS 370Steol CS 370ESteol CS 460Steol DES 32Stepanol CS 230Stepanol CS 460Sulfochem ES 1KSulfochem ES 2Sulfochem ES 2CWKSulfochem ES 2KSulfochem ES 2PKSulfochem ES 3Sulfochem ES 70, Sulfotex 6040X, Sunnol CM 1470, Sunnol DL 1430, Sunnol LMT 1430, Supamin C, Supamin SA, TH 330M, TH 370N, Taipol NLES 227, Taipol NLES 327, Taipol NOES 340, Taycapol NE 1230, Taycapol NE 7030, Tensaryl SBU, Texapon 15, Ultrasulfate SE 5, Ungerol LES 3-70, Witcolate ES 2, Witcolate ES 3, Witcolate ES 370, Witcolate LES 60C, Witcolate S 1285C, Zetesol 250, Zetesol 270, Zetesol LES 2, Zetesol LES 2A, Zetesol LES 3SL, Zetesol NL, Zetesol NL-2, Polystep B 12, Polystep B 19, Polystep B 23, RSWA-ESB 70ZA, Retzolate 60, Rewopol NL 2, Rewopol NL 2-28, Rewopol NL 3RhodaPex ESB 70NAT, Rhodapex 3N70, SBL 3N27, SLES 1EO, SLES 2EO, SLES 2EO SMU, SLES 3EO, SLES 3EO SMU, SN, SPE 1150, SPE 1200K, SPE 1250, SPE 1350, Sandet EN, Sapanol DSS, Sapanol VS, Shinoline SPE 1350, Sinolin 1150, Sinolin SPE 1150, Sinolin SPE 1200, Sinolin SPE 1200K, Sinolin SPE 1250, Sinolin SPE 1350, Sipon AOS 225UP, Sipon ES, Sipon ES 2, Sipon ESY, Sipon LES 25, Sipon LES 328, Lauropal 12, MA 101, Manro NEC 28, Maprofix 60S, Maprofix ES, Marlinat 242/28, Montelane KRO, N 25, N 335T, Neolight EXT, Neopon LOS, Neopon LOS 3N70, Neopon LOS 70, Neopon LOS/NF, Nikkol NES 203, Nikkol Phosten SBL 3N27, Nikkol SBL 2N27, Nikkol SBL 3N27, Nikkol SBL 4N, Nissan Persoft EF, Nissan Persoft EP, Nissan Trax K 300, PCA 078, Persoft EF, Persoft EP, Pionin A 28B |
| InChI Formula | InChI=1S/C14H30O5S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-18-13-14-19-20(15,16)17;/h2-14H2,1H3,(H,15,16,17);/q;+1/p-1 |
| SMILES | CCCCCCCCCCCCOCCOS(=O)(O[Na])=O |
| IUPAC Name | Sodium 2-(dodecyloxy)ethyl sulfate |
| Molecular Weight (g/mol) | 332.43 |
| Formula Weight | 332.16 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?