missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium L-Lactate-2-d1, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Sodium L-Lactate-2-D |
| CAS | 285979-83-1 |
| Molecular Formula | C3 D H4 O3 . Na |
| Quantity | 0.05 g |
| Synonym | Propanoic-2-d acid, 2-hydroxy-, monosodium salt, (2 S)- (9 CI), L-(+)-Lactic acid-d1 sodium salt, Sodium S-lactate-2-d, Sodium (S)-2-hydroxypropanoate-2-d, (S)-2-Hydroxypropionic-2-d acid sodium salt, Sodium (S)-lactate-2-d, Sodium L-(+)-lactate-d, Sodium L-Lactate-2-d1 |
| InChI Formula | InChI=1 S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4 H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i2D; |
| SMILES | [Na+].[2 H][C@@](C)(O)C(=O)[O-] |
| IUPAC Name | sodium;(2 S)-2-deuterio-2-hydroxypropanoate |
| Molecular Weight (g/mol) | 113.0659 |
| Formula Weight | 113.0199 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction