missing translation for 'onlineSavingsMsg'
Learn More

Sodium isopropoxide isopropanol adduct, 95%, Thermo Scientific™

Product Code. 15412427
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412427 25 g 25g
15402427 5 g 5g
15422427 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15412427 Supplier Thermo Scientific Alfa Aesar Supplier No. 045648.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula NaOC3H7·C3H8O
MDL Number MFCD12547728

Specifications

Melting Point 150°C (decomposition)
Color Cream
Linear Formula NaOCH(CH3)2·C3H7OH
Quantity 25 g
UN Number UN3206
Sensitivity Air & Moisture Sensitive
Formula Weight 142.17
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Sodium isopropoxide isopropanol adduct
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.