missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 32% weight Solution, Chem-Lab

Product Code. 13288303
Click to view available options
Quantity:
2.5 L
Packaging:
PE
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 13288303

Brand: Chem-Lab CL02.4002.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pure solution for laboratory use.

Specifications

CAS 1310-73-2
Molecular Formula HNaO
Concentration 432g NaOH / 1L H2O
UN Number UN1824
Density 1.349g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 2.5 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Percent Purity 32% (weight)
Packaging PE
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.