Learn More
Sodium Hydrosulfite (85%), TRC
Product Code. 30416575
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416575 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416575 Supplier Toronto Research Chemicals Supplier No. TRCS63481025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium Dithionite
CAS 7775-14-6
Molecular Formula 2 Na . O4 S2
Quantity 25 g
Synonym Dithionous acid','sodium salt (1:2), Dithionous acid','disodium salt (8CI,9CI), Blankit, Blankit IN, Blankit S, Burmol, Cyclanon ARC, Disodium dithionite, Disodium hydrosulfite, Hydros, Hydrosulfite R Conc, Sodium dithionite, Sodium dithionite (Na2(S2O4)), Sodium hydrosulfite, Sodium hydrosulfite (Na2S2O4), Sodium hyposulfite, V-Brite B, Vatrolite
InChI Formula InChI=1S/2Na.H2O4S2/c;;1-5(2)6(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2
SMILES [Na+].[Na+].[O-]S(=O)S(=O)[O-]
Molecular Weight (g/mol) 174.11
Formula Weight 173.9033