Learn More
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Product Code. 11375045
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375045 1000 g 1000g
11385045 250 g 250g
15405095 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11375045 Supplier Thermo Scientific Alfa Aesar Supplier No. B25587.0B
Request Bulk or Custom Format
CAS: 7681-38-1 | HNaO4S | 120.05 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
SMILES [Na+].OS([O-])(=O)=O
Specifications
Melting Point 177°C to 180°C
pH 1.0
Quantity 1000 g
UN Number UN3260
Merck Index 14,8586
IUPAC Name sodium;hydrogen sulfate
Formula Weight 120.06
Percent Purity ≥90%
Odor Odorless
Sensitivity Hygroscopic
Density 2.435
Assay remainder mainly sodium sulfate
Chemical Name or Material Sodium hydrogen sulfate, anhydrous