missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen phosphate, 98+%

Product Code. 11448970
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
10 kg
Unit Size:
10000g
1kg
2500g
250g
500g
5kg
This item is not returnable. View return policy

Product Code. 11448970

Brand: Thermo Scientific Alfa Aesar A11817.A7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol

Sodium hydrogen phosphate is used in food and water treatment in conjunction with trisodium phosphate. It is used as an emulsifier and buffer in foods and pharmaceuticals, anti-caking additive in powdered products, dietary supplement, scale inhibitor, reagent and buffer in analytical chemistry. It finds application in detergents, cleaning agents, textile, leather auxiliary, fertilizers and galvanoplastics. Further, it is used as a saline laxative to treat constipation. It inhibits the calcium scale formation in water treatment plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen phosphate is used in food and water treatment in conjunction with trisodium phosphate. It is used as an emulsifier and buffer in foods and pharmaceuticals, anti-caking additive in powdered products, dietary supplement, scale inhibitor, reagent and buffer in analytical chemistry. It finds application in detergents, cleaning agents, textile, leather auxiliary, fertilizers and galvanoplastics. Further, it is used as a saline laxative to treat constipation. It inhibits the calcium scale formation in water treatment plants.

Solubility
Soluble in water. Insoluble in ethanol, methanol and n-octanol.

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 5 kg
Linear Formula Na2HPO4
Merck Index 14,8659
Solubility Information Soluble in water. Insoluble in ethanol,methanol and n-octanol.
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.98
Percent Purity ≥98%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Sodium hydrogen phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.