missing translation for 'onlineSavingsMsg'
Learn More

Sodium fluoride, Honeywell™

Product Code. 15666790 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15666790 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15666790 Supplier Honeywell Research Chemicals Supplier No. 715221KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Purum p.a.,≥98.0% (F)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7681-49-4
Quantity 1 kg
Molecular Formula FNa
Linear Formula NaF
MDL Number MFCD00003524
UN Number UN1690
Synonym sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES [F-].[Na+]
IUPAC Name sodium fluoride
Molecular Weight (g/mol) 41.99
PubChem CID 5235
ChEBI CHEBI:28741
Formula Weight 41.99g/mol
Percent Purity ≥98% (F)
Vapor Pressure 1.4 mmHg (0°C)
Chemical Name or Material Sodium fluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.