Learn More
Sodium Ethyl Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl sulfate sodium salt (as ethyl sulfate) |
| CAS | 546-74-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C2 H5 O4 S . Na |
| Quantity | 1 g |
| Synonym | Sulfuric acid','monoethyl ester','sodium salt (1:1), Ethyl sodium sulfate (6CI,7CI), Sulfuric acid','monoethyl ester','sodium salt (8CI,9CI), Sodium ethyl sulfate, Sodium sulfovinate, Ethyl Sulfate Sodium Salt |
| InChI Formula | InChI=1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1 |
| SMILES | [Na+].CCOS(=O)(=O)[O-] |
| IUPAC Name | sodium;ethyl sulfate |
| Molecular Weight (g/mol) | 148.11 |
| Show More |