Learn More
Sodium Dodecylsulfate, ≥98%, Ultrapure
Product Code. 15805228 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
Product Code. 15805228
Brand: Thermo Scientific J75819.22
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium;dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Specifications
Physical Form Crystalline Flakes
Melting Point ∼206°C
Quantity 100 g
Packaging Plastic bottle
Linear Formula CH3(CH2)11OSO3Na
UN Number UN2926
Beilstein 3599286
Formula Weight 288.38
Percent Purity ≥98%
Grade Ultra Pure
Chemical Name or Material Sodium Dodecylsulfate