Learn More
Sodium deuteroxide, for NMR, 40 wt% solution in D2O, 99+ atom % D
Product Code. 10377830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10377830 50 g Plastic Bottle 50g
10573694 10 g Plastic Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10377830 Supplier Thermo Scientific Acros Supplier No. 175470500
  • ONLINE EXCLUSIVE PRICE
    141.00€
    136.00€ / 50g
    Save 5.00€
    3% Off
Estimated Shipment: 15-07-2026
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 14014-06-3 | HNaO | 41.003 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14014-06-3 , 7789-20-0
Molecular Formula HNaO
Molecular Weight (g/mol) 41.003
MDL Number MFCD00037669
InChI Key HEMHJVSKTPXQMS-DYCDLGHISA-M
Synonym sodium deuteroxide, sodium hydroxide na od, sodium deuteroxide, 40 wt% in deuterium oxide, naod, sodium hydroxide-d, sodium deuteroxide solution, sodium deuteroxide, 1 m in deuterium oxide, sodium deuteroxide, 0.1 m in deuterium oxide, sodium deuteroxide, 30 wt% in deuterium oxide, sodium hydroxide na od 6ci,7ci,8ci,9ci
PubChem CID 23676750
SMILES [OH-].[Na+]
Specifications
Physical Form Liquid
Color Colorless
Density 1.4600g/mL
Deuterium (D) 99.0% min.
Packaging Plastic Bottle
Refractive Index 1.4039
Quantity 50 g
Linear Formula NaOD
Formula Weight 41
Grade NMR
Chemical Name or Material Sodium deuteroxide, 99+ atom % D
Show More Show Less
compliance-icons
Product Identifier
  • Sodium deuteroxide, 30 wt% solution in D2O
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.