missing translation for 'onlineSavingsMsg'
Learn More

Sodium Dehydroepiandrosterone-16,16-d2 Sulfate, CDN

Product Code. 30314322
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30314322

Brand: CDN CDND52970.005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Prasterone-16,16-D2 Sulfate
CAS 352431-52-8
Molecular Formula C19 D2 H25 O5 S . Na
Quantity 0.005 g
Synonym Androst-5-en-17-one-16,16-d2, 3-(sulfooxy)-, sodium salt, (3β)- (9 CI), (3β)-3-(Sulfooxy)androst-5-en-17-one-16,16-d2 sodium salt, Dehydroepiandrosterone-16,16-d2 sulfate sodium salt, Prasterone-16,16-d2 sodium sulfate, Sodium dehydroepiandrosterone-16,16-d2 sulfate, Sodium dehydroepiandrosterone-3-sulfate-16,16-d2, Sodium prasterone-16,16-d2 sulfate
InChI Formula InChI=1 S/C19H28O5S.Na/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;/h3,13-16 H,4-11H2,1-2H3,(H,21,22,23);/q;+1/p-1/t13-,14-,15-,16-,18-,19-;/m0./s1/i6D2;
SMILES [Na+].[2 H]C1([2 H])C[C@H]2 [C@@H]3 CC=C4C[C@H](CC[C@]4(C)[C@H]3 CC[C@]2(C)C1=O)OS(=O)(=O)[O-]
IUPAC Name sodium;[(3 S,8 R,9 S,10 R,13 S,14 S)-16,16-dideuterio-10,13-dimethyl-17-oxo-2,3,4,7,8,9,11,12,14,15-decahydro-1 H-cyclopenta[a]phenanthren-3-yl] sulfate
Molecular Weight (g/mol) 392.4818
Formula Weight 392.1602 g/mol
Percent Purity 97 atom % D, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.