missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate-2,2,4,4-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Trisodium Citrate-2,2,4,4-D4 |
| Molecular Formula | C6 D4 H O7 . 3 Na |
| Quantity | 0.1 g |
| Synonym | Trisodium citrate-2,2,4,4-d4, Citric-2,2,4,4-d4 acid sodium salt (1:3), Sodium citrate-2,2,4,4-d4, 2-Hydroxy-1,2,3-propane-1,1,3,3-d4-tricarboxylic acid trisodium salt |
| InChI Formula | InChI=1 S/C6H8O7.3 Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3/i1D2,2D2;;; |
| SMILES | [Na+].[Na+].[Na+].[2 H]C([2 H])(C(=O)[O-])C(O)(C(=O)[O-])C([2 H])([2 H])C(=O)[O-] |
| IUPAC Name | trisodium;1,1,3,3-tetradeuterio-2-hydroxypropane-1,2,3-tricarboxylate |
| Molecular Weight (g/mol) | 262.094 |
| Formula Weight | 261.998 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction