Learn More
Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm, polished both sides
Product Code. 15476245
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 15476245
Brand: Thermo Scientific Alfa Aesar 038455.KT
Request Bulk or Custom Format
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Spectroscopy, IR window
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7647-14-5
Quantity 1 Ea.
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium;chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Chemical Name or Material Sodium chloride crystal optic rectangle, 30mm x 15mm x 4mm, Polished both sides