Learn More
Sodium bisulfate, Honeywell Fluka™
Product Code. 15676810 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15676810 5 kg 5kg
15666810 250 g 250g
15656810 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15676810 Supplier Honeywell-Fluka Supplier No. 716565KG
Request Bulk or Custom Format
Purum, anhydrous, ∼95.0%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
IUPAC Name sodium;hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O
Specifications
Quantity 5 kg
Linear Formula NaHSO4
UN Number NONH for all modes of transport
Formula Weight 120.06g/mol
Percent Purity 93 to 102% (T), 95%
Chemical Name or Material Sodium bisulfate
compliance-icons
Product Identifier
  • Sodium bisulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydrogensulphate