Learn More
R&DSystems™ Sodium Bicarbonate, powder
Product Code. 18431970
Click to view available options
Quantity:
500 g
1 Kg
Unit Size:
1kg
500g
Product Code. 18431970
Brand: R&DSystems™ R39150
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Cell culture grade sodium bicarbonate in powder form.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature, under dry conditions
For Use With (Application) Cell Culture
Quantity 500 g