Learn More
Sodium 6-hydroxy-5-((2-methoxy-5-methyl-4-sulfonatophenyl)diazenyl)naphthalene-2-sulfonate, 85%, BLD Pharm™
Product Code. 30948378
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948378 500 g 500g
30943007 100 g 100g
30938875 25 g 25g
30936785 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30948378 Supplier BLD Pharm Supplier No. BD113032500g
 Restricted Item
Estimated Shipment: 27-11-2026
Specifications
CAS 25956-17-6
Molecular Formula C18H14N2Na2O8S2
MDL Number MFCD00059526
Quantity 500 g
SMILES O=S(C1=CC=C2C(/N=N/C3=CC(C)=C(S(=O)([O-])=O)C=C3OC)=C(O)C=CC2=C1)([O-])=O.[Na+].[Na+]
Molecular Weight (g/mol) 496.42
Percent Purity 85%
Chemical Name or Material Sodium 6-hydroxy-5-((2-methoxy-5-methyl-4-sulfonatophenyl)diazenyl)naphthalene-2-sulfonate