Learn More
Sodium 3-(diphenylphosphino)benzenesulfonate, 90%, BLD Pharm™
Specifications
Specifications
| CAS | 63995-75-5 |
| Molecular Formula | C18H14NaO3PS |
| MDL Number | MFCD00144157 |
| Quantity | 250 mg |
| SMILES | O=S(C1=CC(P(C2=CC=CC=C2)C3=CC=CC=C3)=CC=C1)([O-])=O.[Na+] |
| Molecular Weight (g/mol) | 364.33 |
| Percent Purity | 90% |
| Chemical Name or Material | Sodium 3-(diphenylphosphino)benzenesulfonate |