missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-Ketobutyrate-3,3,4,4,4-d5, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Sodium 2-Oxobutanoate-D5 |
| Molecular Formula | C4 D5 O3 . Na |
| Quantity | 0.05 g |
| Synonym | 2-Oxobutanoic-3,3,4,4,4-d5 acid sodium salt, 2-Oxobutanoic-D5 acid sodium salt, 3-Methylpyruvic-3,3,4,4,4-d5 acid sodium salt, α-Ketobutyric-β-d2 acid sodium salt, Sodium 2-oxobutanoate-D5, Sodium 3-methylpyruvate-d5, Sodium 2-ketobutyrate-3,3,4,4,4-d5 |
| InChI Formula | InChI=1 S/C4H6O3.Na/c1-2-3(5)4(6)7;/h2H2,1H3,(H,6,7);/q;+1/p-1/i1D3,2D2; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)C(=O)[O-] |
| IUPAC Name | sodium;3,3,4,4,4-pentadeuterio-2-oxobutanoate |
| Molecular Weight (g/mol) | 129.101 |
| Formula Weight | 129.045 g/mol |
| Percent Purity | 98 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction