missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-ethylhexanoate, 97%

Product Code. 11401507
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11401507

Brand: Thermo Scientific Alfa Aesar B24960.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19766-89-3 | C8H15NaO2 | 166.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19766-89-3
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD00014007
InChI Key VYPDUQYOLCLEGS-UHFFFAOYNA-M
Synonym sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate
PubChem CID 23672331
IUPAC Name sodium;2-ethylhexanoate
SMILES [Na+].CCCCC(CC)C([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 1000 g
Solubility Information Soluble in water.
Formula Weight 166.2
Percent Purity 97%
Chemical Name or Material Sodium 2-ethylhexanoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.