missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%

Product Code. 11480431
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11480431

Brand: Thermo Scientific Alfa Aesar A19995.06

This item is not returnable. View return policy

CAS: 15484-44-3 | C2H4ClNaO3S | 166.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15484-44-3
Molecular Formula C2H4ClNaO3S
Molecular Weight (g/mol) 166.55
MDL Number MFCD00149546 MFCD00007531
InChI Key BVIXLMYIFZGRBH-UHFFFAOYSA-M
Synonym sodium 2-chloroethylsulfonate, 2-chloroethane sulfonic acid sodium salt, sodium .beta.-chloroethanesulfonate, sodium 2-chloroethane-1-sulfonic acid, sodium 2-chloroethanesulfonic acid
PubChem CID 54611855
SMILES [Na+].[O-]S(=O)(=O)CCCl

Specifications

Melting Point ∼292°C (decomposition)
Quantity 5 g
Beilstein 5649202
Sensitivity Hygroscopic
IUPAC Name sodium;2-chloroethanesulfonic acid
Formula Weight 166.56 (Anhydrous)
Percent Purity ≥98%
Assay (dry wt.), water <10%
Chemical Name or Material Sodium 2-chloroethanesulfonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.