Learn More
Rogo Sampaic™ Soda Lime Glass Test Tubes
Product Code. 11835953
Click to view available options
Diameter (Metric):
15.3 mm
17.3 mm
Length (Metric):
160 mm
180 mm
Unit Size:
Pack of 250
Pack of 500
Product Code. 11835953
Brand: Rogo Sampaic™ BGB030
Ideal for routine laboratory analysis and procedures. Rogo Sampaic™ Soda Lime Glass Test Tubes are economical while still offering good chemical resistance.
  • Thin wall
  • Available in two different capacities
Specifications
Description Test tube
Length (Metric) 160 mm
Material Soda Lime Glass
Quantity 500 Pack
Diameter (Metric) 15.3 mm