Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Product Code. p-8003884
Change view
Click to view available options
Length (Metric):
40 mm
50 mm
55 mm
70 mm
120 mm
125 mm
150 mm
Diameter (Metric):
8 mm
9.5 mm
10.5 mm
11 mm
12 mm
16 mm
Unit Size:
Pack of 200
Pack of 250
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11882393 40 mm 10.5 mm Pack of 500
11892393 55 mm 11 mm Pack of 500
11842383 70 mm 11 mm Pack of 200
11803370 120 mm 12 mm Pack of 500
11862393 125 mm 16 mm Pack of 250
11832383 50 mm 9.5 mm Pack of 200
11822383 40 mm 8 mm Pack of 200
10364713 150 mm 16 mm Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11882393 Supplier DWK Life Sciences Supplier No. A404010500911
Add to basket
Add to basket
Use the Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Design Type Round bottom
Length (Metric) 40 mm
Packaging 500Pack
Diameter (Metric) 10.5 mm
Product Type Test Tube