Learn More
Fisherbrand™ Soda Lime Glass Culture Tube
Use the Fisherbrand™ Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
Brand:  Fisherbrand™ A415018000911
Product Code. 12885443
  • 141.00€ / Pack of 500
Only null left
Estimated Shipment: 03-02-2026
Description
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable