Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Product Code. 15882265 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
100 mm
150 mm
Diameter (Metric):
12 mm
16 mm
Unit Size:
Pack of 100
Product Code. 15882265
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
  • Soda Lime Glass
  • Rimless
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 4142
Length (Metric) 150 mm
Material Soda Lime Glass
Quantity 100 Pack
Rimmed No
Diameter (Metric) 16 mm