Learn More
Brady™ Snap-On™ Pipe Cover
Product Code. 12779884
Change view
Click to view available options
Length (Metric):
228 mm
300 mm
420 mm
Width (Metric):
200 mm
300 mm
420 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) unitSize
12779884 228 mm 200 mm Each
12789884 300 mm 300 mm Each
12799884 420 mm 420 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12779884 Supplier Brady™ Supplier No. 275240
  • ONLINE EXCLUSIVE PRICE
    20.05€
    16.94€ / Pack of 1
    Save 3.11€
    15% Off
In Stock
Add to basket
Add to basket
Snap-On™ Markers is a fast and easy way to mark rusty, wet or oily pipes. The Snap-On™ Pipe Cover is a transparent PVC cover on which you can apply any of the markers in our ranges. The result is that your self-adhesive marker remains readable and well protected.
  • Keeps your self-adhesive markers readable and well protected.
Specifications
Certifications/Compliance Meet ANSI A13.1 requirements
Description Brady Snap-On Pipe Markers
Color Transparent
Finish Gloss
Material Plastic
Product Type Pipe Marker
For Use With (Application) Pipe and valve marking
Labels per Sheet 1
No. per Pack 1 Ea.
Quantity 1 Ea.
Temperature Range (English) -40°F to +170°F
Temperature Range (Metric) -40°C to +77°C
Thickness (English) 0.030 in.
Thickness (Metric) 0.762 mm
Dimensions (L x W) 228 x 200 mm
Dimensions (L x W x H) 228 x 200 x 26 mm
Length (English) 8.98 in.
Length (Metric) 228 mm
Width (English) 7.87 in.
Width (Metric) 200 mm
Height (English) 1.02 in.
Height (Metric) 26 mm
Alternate Part Number SNAP-ON-1-B817