Learn More
behr Labor-Technik™ SMX30 Behrotest™ Workstation for Soxhlet Extraction
Product Code. 10151992
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10151992 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10151992 Supplier behr Labor-Technik™ Supplier No. B00218422
  • ONLINE EXCLUSIVE PRICE
    10589.25€
    7436.31€ / Pack of 1
    Save 3152.94€
    29% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
Accessory for Soxhlet extraction system.
  • Used in solvent extraction process
Specifications
For Use With (Equipment) Soxhlet Extraction System
Product Type Workstation
Capacity (Metric) 30 mL
Includes Six sample holders
For Use With (Application) Work bench for extraction of solvents