missing translation for 'onlineSavingsMsg'
Learn More

Techsit™ Smartsit High Laboratory Chair

Product Code. p-7227799
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Blue
Gray
Red
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17773399 Black Each
17783399 Blue Each
17793399 Gray Each
17703409 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17773399 Supplier Techsit™ Supplier No. TS6002Black

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This antimicrobial laboratory chair boasts a foot ring on glides, and is suitable for bench height.

  • Easy to clean
  • Adjustable foot ring on glides
  • Hard wearing
  • Ergonomically designed and contoured seat and back

Specifications

Color Black
Adjustable (Height) Yes
Back Style Ergonomic
Certifications/Compliance Cleanroom Class 5
Finish Polyurethane
Frame Material Steel
Seating Height 590 to 830 mm
Warranty 5 Year
Weight (English) 15 kg
Wheel Type Glide
Width (Metric) 480 mm
With Armrest No
With Backrest Yes
With Casters No
Footrest Yes
With Lumbar Support Yes
With Tilt No
Seating Type High Bench Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.