Learn More
STEMCELL Technologies SmartDish™ - 50 Pack
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
Description
- SmartDish™ plates enable accurate and reproducible hematopoietic colony counting by preventing the formation of a meniscus
- This allows for an even distribution of culture medium, resulting in a more uniform distribution of colonies throughout the entire 35 mm well
- The absence of the meniscus also reduces optical distortion so that colonies located at the edge can be more easily counted
- SmartDish™ (Catalog #27370) is the second generation of the original SmartDish™ plate (Catalog #27301)
- These new plates use an improved coating, which streamlines the manufacturing process while consistently improving the visibility of hematopoietic colonies in culture through elimination of the mensicus
- Current SmartDish™ customers can contact their local STEMCELL representative for more information
- SmartDish™ has been designed for both manual counting with STEMgrid™-6 as well as automated colony counting with a STEMvision™ instrument
Specifications
Specifications
| For Use With (Application) | Cell Culture; Colony Assay |
| Quantity | 50/Pk. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.