Learn More
STEMCELL Technologies SmartDish™ - 50 Pack
Product Code. 15791668
Encompass_Preferred
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
Product Code. 15791668
Brand: STEMCELL Technologies 27371
Meniscus-free cultureware for more accurate counting of hematopoietic colonies in CFU assays
  • SmartDish™ plates enable accurate and reproducible hematopoietic colony counting by preventing the formation of a meniscus
  • This allows for an even distribution of culture medium, resulting in a more uniform distribution of colonies throughout the entire 35 mm well
  • The absence of the meniscus also reduces optical distortion so that colonies located at the edge can be more easily counted
  • SmartDish™ (Catalog #27370) is the second generation of the original SmartDish™ plate (Catalog #27301)
  • These new plates use an improved coating, which streamlines the manufacturing process while consistently improving the visibility of hematopoietic colonies in culture through elimination of the mensicus
  • Current SmartDish™ customers can contact their local STEMCELL representative for more information
  • SmartDish™ has been designed for both manual counting with STEMgrid™-6 as well as automated colony counting with a STEMvision™ instrument
Specifications
For Use With (Application) Cell Culture; Colony Assay
Quantity 50/Pk.