Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A
Product Code. 15820182 Shop All Thermo Scientific Products
Click to view available options
Description:
Magnetic, Soluble
Magnetic, Standalone
Magnetic, with SOLAμ Collection Plate, Soluble
Non-Magnetic, Soluble
Non-Magnetic, with SOLAμ Collection Plate, Soluble
Unit Size:
Each
Product Code. 15820182
Brand: Thermo Scientific™ 60114104
Use this kit to for purification of immunoglobulins prior to protein A digestion.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Description Magnetic, Soluble
Capture Protein A
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Magnetic
For Use With (Application) Immunocapture and Protein Digest Sample Preparation
Product Line SMART Digest
Type Bulk Fractionation Kit
Includes Bulk Magnetic SMART Digest Soluble Resin